(-)-Epicatechin gallate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-3-yl 3,4,5-trihydroxybenzoate, (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate
A336586-100MG
A336586-10MG
A336586-1MG
A336586-250MG
A336586-25MG
A336586-5MG
A336586-50MG
A336586-1G
A336586-5G
76724-258EA
114
USD
76724-258
76724-260
76724-262
76724-264
76724-266
76724-268
77601-520
77879-724
77879-725
(-)-Epicatechin gallate 98+%
(-)-Epicatechin gallate
Precaución: for Research Use Only
Formula:
C₂₂H₁₈O₁₀ MW: 442.38 g/mol |
MDL Number:
MFCD00075936 CAS Number: 1257-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...