6-Methoxyquinoline-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quininic acid

A336526-100MG A336526-1G A336526-250MG
76724-252EA 71.09 USD
76724-252 76724-254 76724-256
6-Methoxyquinoline-4-carboxylic acid 98%
6-Methoxyquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.20 g/mol
MDL Number: MFCD00024013
CAS Number: 86-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR