N-(Trimethylsilyl)morpholine 97% (by GC)

Supplier: Ambeed
Danger

A336095-5G A336095-25G
76724-246EA 21 USD
76724-246 77668-540
N-(Trimethylsilyl)morpholine 97% (by GC)
N-(Trimethylsilyl)morpholine

Precaución: for Research Use Only

Formula: C₇H₁₇NOSi
MDL Number: MFCD00010098
CAS Number: 13368-42-8
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR