4-Phthalimidobutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1,3-Dioxoisoindolin-2-yl)butanoic acid, 4-(Phthalimid-1-yl)butanoic acid

A335022-100MG A335022-1G A335022-250MG A335022-5G
76724-218EA 17 USD
76724-218 76724-220 76724-222 76724-224
4-Phthalimidobutyric acid 98%
4-Phthalimidobutyric acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₄
MDL Number: MFCD00196079
CAS Number: 3130-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR