Methyl-3-(4-pyridyl)acrylate 95%

Supplier: Ambeed
Warning

A334325-100MG A334325-1G A334325-250MG A334325-5G
76724-194EA 29 USD
76724-194 76724-196 76724-198 76724-200
Methyl-3-(4-pyridyl)acrylate 95%
Methyl-3-(4-pyridyl)acrylate

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00023636
CAS Number: 7340-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR