Dibenzyl phthalate 95%

Supplier: Ambeed
Warning

A333402-250MG A333402-1G A333402-5G
76724-192EA 28 USD
76724-192 77879-712 77879-713
Dibenzyl phthalate 95%
Dibenzyl phthalate

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₄
MDL Number: MFCD00014437
CAS Number: 523-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR