Perylene 95%

Supplier: Ambeed
Danger

A332515-1G A332515-25G A332515-5G
76724-186EA 5.31 USD
76724-186 76724-188 76724-190
Perylene 95%
Perylene

Precaución: for Research Use Only

Formula: C₂₀H₁₂
MW: 252.32 g/mol
MDL Number: MFCD00004142
CAS Number: 198-55-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR