N,N'-Bis(2-hydroxyethyl)oxamide 99%
Supplier: Ambeed
|
Danger
|
A332346-5G
A332346-1G
76724-184EA
26
USD
76724-184
77879-708
N,N'-Bis(2-hydroxyethyl)oxamide 99%
N,N'-Bis(2-hydroxyethyl)oxamide
Precaución: for Research Use Only
Formula:
C₆H₁₂N₂O₄ |
MDL Number:
MFCD00020559 CAS Number: 1871-89-2 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...