Sodium cyclamate 95%

Supplier: Ambeed
Warning

A331698-1000G A331698-100G A331698-25G A331698-500G
76724-176EA 38 USD
76724-176 76724-178 76724-180 76724-182
Sodium cyclamate 95%
Sodium cyclamate

Precaución: for Research Use Only

Formula: C₆H₁₂NNaO₃S
MDL Number: MFCD00003827
CAS Number: 139-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR