2-Propoxyphenol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-2-propoxybenzene

A331040-1G A331040-25G A331040-5G
76724-162EA 18 USD
76724-162 76724-164 76724-166
2-Propoxyphenol 97%
2-Propoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD06655033
CAS Number: 6280-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR