Pyridinium trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridinium Triflate
A330980-100G
A330980-1G
A330980-25G
A330980-5G
76724-154EA
199
USD
76724-154
76724-156
76724-158
76724-160
Pyridinium trifluoromethanesulfonate 98%
Pyridinium trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₆H₆F₃NO₃S MW: 229.17 g/mol |
MDL Number:
MFCD00274656 CAS Number: 52193-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...