Propargyl benzenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Prop-2-yn-1-yl benzenesulphonate

A330653-100G A330653-10G A330653-25G A330653-5G
76724-146EA 36 USD
76724-146 76724-148 76724-150 76724-152
Propargyl benzenesulfonate 97%
Propargyl benzenesulphonate

Precaución: for Research Use Only

Formula: C₉H₈O₃S
MW: 196.2200 g/mol
MDL Number: MFCD00013459
CAS Number: 6165-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR