5'-Chloro-4'-fluoro-2'-nitroacetanilide 98%

Supplier: Ambeed
Warning

A329967-1G A329967-250MG A329967-25G A329967-5G
76724-138EA 16 USD
76724-138 76724-140 76724-142 76724-144
5'-Chloro-4'-fluoro-2'-nitroacetanilide 98%
5'-Chloro-4'-fluoro-2'-nitroacetanilide

Precaución: for Research Use Only

Formula: C₈H₆ClFN₂O₃
MDL Number: MFCD00024581
CAS Number: 81962-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR