4-(Methylthio)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-(methylthio)phenyl)-1,3,2-dioxaborolane

A329573-1G A329573-250MG A329573-5G
76724-132EA 39.26 USD
76724-132 76724-134 76724-136
4-(Methylthio)phenylboronic acid pinacol ester 98%
4-(Methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₂S
MW: 250.1648 g/mol
MDL Number: MFCD05155222
CAS Number: 190788-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR