Ethyl-3-(pyridin-2-ylamino)propanoate 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Pyridylamino)propionic acid ethyl ester, Ethyl 3-(2-Pyridylamino)propionate, Ethyl 3-(pyridin-2-ylamino)propanoate

A329230-100G A329230-10G A329230-1G A329230-25G A329230-500G A329230-5G
76724-100EA 50.93 USD
76724-100 76724-102 76724-104 76724-106 76724-108 76724-110
Ethyl-3-(pyridin-2-ylamino)propanoate 95%
Ethyl-3-(pyridin-2-ylamino)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.2300 g/mol
MDL Number: MFCD09833621
CAS Number: 103041-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR