Dimethyl quinoline-2,3-dicarboxylate 97%

Supplier: Ambeed
Warning

A328019-1G A328019-250MG
76724-082EA 96.55 USD
76724-082 76724-084
Dimethyl quinoline-2,3-dicarboxylate 97%
Dimethyl quinoline-2,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₄
MW: 245.23 g/mol
MDL Number: MFCD00434841
CAS Number: 17507-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR