Potassium bis(trifluoromethylsulfonyl)imide 97%

Supplier: Ambeed
Danger

Synonyms: Potassium bis(trifluoromethanesulphonyl)imide

A327422-5G A327422-250MG A327422-25G A327422-1G
76724-074EA 55 USD
76724-074 77879-689 77879-690 77879-688
Potassium bis(trifluoromethylsulfonyl)imide 97%
Potassium bis(trifluoromethylsulfonyl)imide

Precaución: for Research Use Only

Formula: C₂F₆KNO₄S₂
MW: 319.234 g/mol
MDL Number: MFCD06200829
CAS Number: 90076-67-8
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR