6-Methoxy-2-naphthylacetic acid 97%
Supplier: Ambeed
|
Warning
|
A326956-100MG
A326956-1G
A326956-250MG
A326956-5G
76724-068EA
30
USD
76724-068
76724-070
76724-072
77879-686
6-Methoxy-2-naphthylacetic acid 97%
6-Methoxy-2-naphthylacetic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₂O₃ |
MDL Number:
MFCD00033105 CAS Number: 23981-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...