N,N'-Di-p-tolylbenzidine 98%

Supplier: Ambeed
Warning

Synonyms: N4,N4'-Di-p-tolyl-[1,1'-biphenyl]-4,4'-diamine, N,N'-Bis(4-methylphenyl)benzidine

A326705-25G A326705-5G
76724-060EA 113 USD
76724-060 76724-062
N,N'-Di-p-tolylbenzidine 98%
N,N'-Di-p-tolylbenzidine

Precaución: for Research Use Only

Formula: C₂₆H₂₄N₂
MDL Number: MFCD09833311
CAS Number: 10311-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR