p-Tolyl isobutyrate 97%

Supplier: Ambeed
Warning

Synonyms: Isobutyric Acid p-Tolyl Ester

A326649-1G A326649-25G A326649-5G
76724-052EA 19.1 USD
76724-052 76724-054 76724-056
p-Tolyl isobutyrate 97%
p-Tolyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00026440
CAS Number: 103-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR