3-(2-Naphthyl)acrylic acid 98%

Supplier: Ambeed
Warning

A325866-1G A325866-25G A325866-5G
76724-046EA 15 USD
76724-046 76724-048 76724-050
3-(2-Naphthyl)acrylic acid 98%
3-(2-Naphthyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MDL Number: MFCD00209666
CAS Number: 51557-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR