1,1,1,3,3-Pentamethyl-3-octyldisiloxane 95%

Supplier: Ambeed
Warning

A325478-1G A325478-250MG A325478-5G
76724-038EA 54.11 USD
76724-038 76724-040 77593-926
1,1,1,3,3-Pentamethyl-3-octyldisiloxane 95%
1,1,1,3,3-Pentamethyl-3-octyldisiloxane

Precaución: for Research Use Only

Formula: C₁₃H₃₂OSi₂
MW: 260.56 g/mol
MDL Number: MFCD01861724
CAS Number: 180006-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR