(S)-(-)-1-(2-Naphthyl)ethanol 98%

Supplier: Ambeed
Warning

A325397-100MG A325397-1G A325397-250MG A325397-5G
76724-032EA 17 USD
76724-032 76724-034 76724-036 77879-681
(S)-(-)-1-(2-Naphthyl)ethanol 98%
(S)-(-)-1-(2-Naphthyl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MDL Number: MFCD00064188
CAS Number: 27544-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR