Nile blue chloride (Basic blue 12), Reagent Grade

Supplier: Ambeed
Warning

Synonyms: Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride

A324556-1G A324556-25G A324556-5G
76724-022EA 9.55 USD
76724-022 76724-024 76724-026
Nile blue chloride (Basic blue 12), Reagent Grade
Nile blue chloride (Basic blue 12)

Precaución: for Research Use Only

Formula: C₂₀H₂₀ClN₃O
MW: 353.85 g/mol
MDL Number: MFCD00011936
CAS Number: 2381-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR