1,3-Bis(4-chlorophenyl)urea 98%

Supplier: Ambeed
Warning

A323790-1G A323790-5G A323790-250MG
76724-010EA 34 USD
76724-010 76724-012 77668-440
1,3-Bis(4-chlorophenyl)urea 98%
1,3-Bis(4-chlorophenyl)urea

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂N₂O
MDL Number: MFCD00018541
CAS Number: 1219-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR