Pyrantel pamoate 97%

Supplier: Ambeed
Warning

Synonyms: Pyrantel embonate

A322375-100G A322375-100MG A322375-10G A322375-1G A322375-25G A322375-5G
76723-994EA 415 USD
76723-994 76723-996 76723-998 76724-000 76724-002 76724-004
Pyrantel pamoate 97%
Pyrantel pamoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂S·C₂₃H₁₆O₆
MW: 594.68 g/mol
MDL Number: MFCD00072037
CAS Number: 22204-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR