Methyl-5-bromo-2-methoxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-2-methoxynicotinate, Methyl 5-bromo-2-methoxy-3-pyridinecarboxylate

A321990-10G A321990-1G A321990-5G
76723-988EA 88 USD
76723-988 76723-990 76723-992
Methyl-5-bromo-2-methoxynicotinate 98%
Methyl-5-bromo-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD08064227
CAS Number: 122433-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR