Allyl-2,2,2-Trichloroacetimidate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Allyl 2,2,2-trichloroacetimidate, 2,2,2-Trichloroacetimidic Acid Allyl Ester
A320878-10G
A320878-1G
A320878-25G
76723-976EA
108.22
USD
76723-976
76723-978
76723-980
Allyl-2,2,2-Trichloroacetimidate 98%
Allyl-2,2,2-Trichloroacetimidate
Precaución: for Research Use Only
Formula:
C₅H₆Cl₃NO MW: 202.46 g/mol |
MDL Number:
MFCD00190710 CAS Number: 51479-73-3 UN: 1760 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...