4-(Methylamino)phenol sulfate 98%

Supplier: Ambeed
Warning

A320159-100G A320159-10G A320159-25G A320159-500G
76723-958EA 53 USD
76723-958 76723-960 76723-962 76723-964
4-(Methylamino)phenol sulfate 98%
4-(Methylamino)phenol sulfate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₆S
MDL Number: MFCD00013140
CAS Number: 55-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR