Phentolamine mesylate 99%

Supplier: Ambeed
Warning

Synonyms: Phentolamine Methanesulphonate, 3-(((4,5-Dihydro-1H-imidazol-2-yl)methyl)(p-tolyl)amino)phenol methanesulphonate, 2-[N-(m-Hydroxyphenyl)-p-toluidinomethyl]imidazoline Methanesulphonate, Phentolamine methanesulphonate salt

A320047-100MG A320047-1MG A320047-250MG A320047-5G A320047-1G
76723-950EA 25.47 USD
76723-950 76723-952 76723-954 76723-956 77668-426
Phentolamine mesylate 99%
Phentolamine mesylate

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O
MW: 377.46 g/mol
MDL Number: MFCD00134201
CAS Number: 65-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR