4-Isopropylcinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Isopropylphenyl)acrylic acid

A319773-1G A319773-25G A319773-5G
76723-944EA 20 USD
76723-944 76723-946 76723-948
4-Isopropylcinnamic acid 98%
4-Isopropylcinnamic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00016544
CAS Number: 3368-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR