Potassium(4-methyl-1-naphthalene)trifluoroborate 96%

Supplier: Ambeed
Warning

A318768-1G A318768-250MG A318768-5G
76723-932EA 28 USD
76723-932 76723-934 76723-936
Potassium(4-methyl-1-naphthalene)trifluoroborate 96%
Potassium (4-methyl-1-naphthalene)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₁H₉BF₃K
MW: 248.09 g/mol
MDL Number: MFCD04115748
CAS Number: 850623-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR