Potassium (4-acetylphenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-acetylphenyltrifluoroborate, Potassium (4-acetylphenyl)trifluoroboranuide

A318488-1G A318488-5G
76723-920EA 14 USD
76723-920 76723-922
Potassium (4-acetylphenyl)trifluoroborate 98%
Potassium 4-acetylphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₇BF₃KO
MW: 226.05 g/mol
MDL Number: MFCD07784376
CAS Number: 252726-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR