9-Chloro-1,2,3,4-tetrahydroacridine 95%

Supplier: Ambeed
Danger

A316529-100MG A316529-1G A316529-5G
76723-898EA 25.47 USD
76723-898 76723-900 76723-902
9-Chloro-1,2,3,4-tetrahydroacridine 95%
9-Chloro-1,2,3,4-tetrahydroacridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN
MDL Number: MFCD00065216
CAS Number: 5396-30-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR