N-[2-(Acetylamino)phenyl]acetamide 97%

Supplier: Ambeed
Warning

A316261-1G A316261-100MG
76723-896EA 80 USD
76723-896 77879-648
N-[2-(Acetylamino)phenyl]acetamide 97%
N-[2-(Acetylamino)phenyl]acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD00092752
CAS Number: 2050-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR