Pyromellitic diimide 97%

Supplier: Ambeed
Warning

Synonyms: Benzene-1,2,4,5-tetracarboxylic Diimide

A316208-1G A316208-5G A316208-25G
76723-892EA 10 USD
76723-892 76723-894 77668-392
Pyromellitic diimide 97%
Pyromellitic diimide

Precaución: for Research Use Only

Formula: C₁₀H₄N₂O₄
MDL Number: MFCD00005004
CAS Number: 2550-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR