4-Formylbenzeneboronic acid, neopentyl glycol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5,5-Dimethyl-2-(4-formylphenyl)-1,3,2-dioxaborinane, 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzaldehyde

A315907-1G A315907-250MG A315907-5G
76723-886EA 12 USD
76723-886 76723-888 76723-890
4-Formylbenzeneboronic acid, neopentyl glycol ester 98%
4-Formylbenzeneboronic acid, neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BO₃
MDL Number: MFCD06796332
CAS Number: 128376-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR