3-(1-Naphthyl)acrylic acid 95%

Supplier: Ambeed
Warning

A315414-10G A315414-1G A315414-25G A315414-5G
76723-870EA 57 USD
76723-870 76723-872 76723-874 76723-876
3-(1-Naphthyl)acrylic acid 95%
3-(1-Naphthyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MDL Number: MFCD00014317
CAS Number: 13026-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR