Sulfadiazine sodium 97%

Supplier: Ambeed
Danger

Synonyms: Sodium sulphadiazine, 4-Amino-N-(pyrimidin-2-yl)benzenesulphonamide, sodium salt

A314813-100G A314813-100MG A314813-10G A314813-25G A314813-500G
76723-860EA 61 USD
76723-860 76723-862 76723-864 76723-866 76723-868
Sulfadiazine sodium 97%
Sulfadiazine sodium

Precaución: for Research Use Only

Formula: C₁₀H₉N₄NaO₂S
MW: 273.2668 g/mol
MDL Number: MFCD00067333
CAS Number: 547-32-0
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR