N-(n-Propyl)maleimide 98+%

Supplier: Ambeed
Danger

Synonyms: 1-Propyl-1H-pyrrole-2,5-dione

A314572-1G A314572-5G A314572-25G
76723-856EA 15 USD
76723-856 76723-858 77879-642
N-(n-Propyl)maleimide 98+%
N-(n-Propyl)maleimide

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.15 g/mol
MDL Number: MFCD00014541
CAS Number: 21746-40-7
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR