Phenylphosphinic acid 98%

Supplier: Ambeed
Danger

A314141-10G A314141-25G
76723-850EA 9 USD
76723-850 76723-852
Phenylphosphinic acid 98%
Phenylphosphinic acid

Precaución: for Research Use Only

Formula: C₆H₇O₂P
MDL Number: MFCD00002131
CAS Number: 1779-48-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR