(3,4-Dimethoxyphenyl)acetic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl (3,4-dimethoxyphenyl)acetate, Ethyl 2-(3,4-dimethoxyphenyl)acetate

A312783-100G A312783-5G A312783-25G
76723-842EA 173.99 USD
76723-842 76723-844 77668-380
(3,4-Dimethoxyphenyl)acetic acid ethyl ester 98%
(3,4-Dimethoxyphenyl)acetic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.26 g/mol
MDL Number: MFCD00017272
CAS Number: 18066-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR