Ethyl-5-methoxyindole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxyindole-2-carboxylic acid ethyl ester

A310663-10G A310663-1G A310663-250MG A310663-5G A310663-25G
76723-818EA 72 USD
76723-818 76723-820 76723-822 76723-824 77879-631
Ethyl-5-methoxyindole-2-carboxylate 98%
Ethyl-5-methoxyindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD00047163
CAS Number: 4792-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR