N-Acetyl-ε-caprolactam 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-2-oxohexamethyleneimine, N-Acetylcaprolactam

A310382-1000G A310382-100G A310382-25G A310382-500G A310382-5G
76723-808EA 238.71 USD
76723-808 76723-810 76723-812 76723-814 76723-816
N-Acetyl-ε-caprolactam 98%
N-Acetyl-epsilon-caprolactam

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
MDL Number: MFCD00003262
CAS Number: 1888-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR