Methyl 2-chloro-2-phenylacetate 98%

Supplier: Ambeed
Danger

A309008-10G A309008-1G A309008-5G A309008-250MG
76723-792EA 175.05 USD
76723-792 76723-794 76723-796 77668-368
Methyl 2-chloro-2-phenylacetate 98%
Methyl 2-chloro-2-phenylacetate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
MDL Number: MFCD00040996
CAS Number: 7476-66-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR