N-Lauroyl-L-glutamic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-Lauroyl-Glu-OH

A308948-1G A308948-250MG
76723-788EA 39.26 USD
76723-788 76723-790
N-Lauroyl-L-glutamic acid 95%
N-Lauroyl-L-glutamic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₁NO₅
MW: 329.43 g/mol
MDL Number: MFCD06410798
CAS Number: 3397-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR