N-(2-Hydroxyethyl)nicotinamide 95%

Supplier: Ambeed
Warning

A307951-10G A307951-1G A307951-250MG A307951-5G
76723-780EA 431 USD
76723-780 76723-782 76723-784 76723-786
N-(2-Hydroxyethyl)nicotinamide 95%
N-(2-Hydroxyethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD00547113
CAS Number: 6265-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR