Sorbitan monolaurate Saponification Value 160-175
Supplier: Ambeed
|
Warning
|
A307256-1000G
A307256-100G
76723-766EA
15
USD
76723-766
76723-768
Sorbitan monolaurate Saponification Value 160-175
Sorbitan monolaurate
Precaución: for Research Use Only
Formula:
C₁₈H₃₄O₆ |
MDL Number:
MFCD00005365 CAS Number: 1338-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...