Pyrilamine maleate 98%

Supplier: Ambeed
Warning

Synonyms: Mepyramine maleate, Anisopyradamine, Minihist, Paraminyl maleate, N1-(4-Methoxybenzyl)-N2,N2-dimethyl-N1-(pyridin-2-yl)ethane-1,2-diamine maleate, Diaminide maleate

A305915-100MG A305915-250MG
76723-714EA 5.31 USD
76723-714 76723-716
Pyrilamine maleate 98%
Pyrilamine maleate

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₃O₅
MW: 401.463 g/mol
MDL Number: MFCD00069333
CAS Number: 59-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR