N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Cyano-3-(trifluoromethyl)phenyl)methacrylamide

A305606-100G A305606-10G A305606-25G A305606-500G A305606-5G
76723-694EA 62 USD
76723-694 76723-696 76723-698 76723-700 76723-702
N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide 98%
N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide

Precaución: for Research Use Only

Formula: C₁₂H₉F₃N₂O
MDL Number: MFCD03411609
CAS Number: 90357-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR